Få mere at vide
Cole-Parmer™ Standard Digital Gloss Meter
Artikelnummer. 13476503 Shop alle Cole-Parmer produkter
Skift visning
Voltage: 1.5V
Artikelnummer.
13476503 Voltage: 1.5V
1 options
Artikelnummer. 13476503 Leverandør Cole-Parmer™ Leverandørnr. 5960005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Use this gloss meter to evaluate the gloss of any coated surface, check paints or inks, inspect print materials, and more.
  • Using an infrared source that emits a beam of light at an angle to the surface, a sensor measures the reflected light.
  • Features a 3-digit LCD and is switch selectable for a standard 60° measurement angle or a 20° angle for measuring high-gloss surfaces.
  • The sensor is separate from the meter for measuring hard-to-reach surfaces.
  • Includes 1-ft (30.5 cm) coiled connection cable and four 1.5 V AAA batteries.