Learn More
Bochem™ Stainless Steel Universal Scissors With Plastic Handle
Product Code. 10303951
Change view
Click to view available options
Length (Metric):
190 mm
210 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10303951 190 mm Each
10498242 210 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10303951 Supplier Bochem™ Supplier No. 4250
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Forventet forsendelse: 22-12-2026
Fashioned with ergonomic handle. Bochem™ Stainless Steel Universal Scissors With Plastic Handle have a stainless steel blade.
  • Plastic handle outside
  • Persistant metal handle
  • Stainless steel blade
Specifications
Product Type Universal Scissors
Length (Metric) 190 mm
Material Stainless Steel