Learn More
Cole-Parmer™ Stainless Steel Tweezers with Flat Tip
Product Code. 11765449 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
12.0 cm
Unit Size:
Each
Product Code. Length (Metric) Unit Size
11765449 12.0 cm Each
1 options
Product Code. 11765449 Supplier Cole-Parmer™ Supplier No. 0728705
  • ONLINE EXCLUSIVE PRICE
    125.00 DKK
    88.61 DKK / Pakke med 1
    Spare 36.39 DKK
    29% Slukket
Forventet forsendelse: 29-12-2026
Recommended for micro electronics and precision assembly
  • Finely crafted stainless steel tweezers
  • Feature hand finished points for accuracy and excellent tip hardness
  • Anti-glare finish reduces eye strain
  • Anti-magnetic, anti-acid and acid resistant
Specifications
Product Type Tweezers
Length (English) 4.75 in.
Length (Metric) 12.0 cm
Material Stainless Steel
Quantity Ea.
Sterility Sterile
Tip Style Tapered, Flat