Få mere at vide
Amsta Rostfritt Stainless Steel Transport Container Without Skirtband, With Lid
Kapacitet (metrisk):
20 l
25 l
30 l
50 l
75 l
100 l
Beskrivelse
- Stainless steel 304 /1.4301
- Straight cylindrical vessel in highly polished finish with stylish design.
Tekniske data
Tekniske data
| Produkttype | Pail |
| Kapacitet (metrisk) | 100 l |
| Farve | Silver |
| Materiale | Stainless Steel |