Learn More
Sartorius™ Stainless Steel OIML Class F2 Reference Line Weight
Stainless Steel OIML Class F2 Reference Line Weight without DAkkS Calibration Certificate.
Brand: Sartorius™ YCW654-00
5924.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Without DAkkS Calibration Certificate
Specifications
Stainless Steel | |
Wood | |
5 kg |
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F2 Reference Line Weight > Wood; Mass: 5 kg
Spot an opportunity for improvement?Share a Content Correction