Learn More
Sartorius™ Stainless Steel OIML Class F1 Reference Line Weight
Stainless Steel OIML Class F1 Reference Line Weight without DAkkS Calibration Certificate.
Brand:  Sartorius™ YCW513-00
1313.13 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10479881
  • 1350.00 DKK / Each
Only null left
Estimated Shipment: 08-07-2025
Description
Stainless Steel OIML Class F1 Reference Line Weight without DAkkS Calibration Certificate.
  • Without DAkkS Calibration Certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
Wood
100 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F1 Reference Line Weight > Wood; Mass: 100 g