Learn More
Sartorius™ Stainless Steel OIML Class F1 Calibration Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW6138-02
2693.41 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10712485
  • 2765.00 DKK / Each
Only null left
Estimated Shipment: 07-07-2025
Specifications
Plastic
1 kg
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F1 Calibration Weight With DAkkS Calibration Certificate > Mass: 1kg