Learn More
Sartorius™ Stainless Steel OIML Class F1 Calibration Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW3238-02
1074.38 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10620086
  • 1105.00 DKK / Each
Estimated Shipment: 11-07-2025
Specifications
Plastic
2 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F1 Calibration Weight With DAkkS Calibration Certificate > Mass: 2g