Learn More
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW622-02
6908.85 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10576577
  • 7085.00 DKK / Each
Only null left
Estimated Shipment: 14-08-2025
Specifications
Wood
2 kg
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight With DAkkS Calibration Certificate > Mass: 2kg