Learn More
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW452-02
1790.63 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10167341
  • 1840.00 DKK / Each
Only null left
Estimated Shipment: 11-08-2025
Specifications
Wood
50 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight With DAkkS Calibration Certificate > Mass: 50g