Learn More
Amsta Rostfritt Stainless Steel Lid For 608/609-150
Product Code. 30284921
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
30284921 None Each
1 options
Product Code. 30284921 Supplier Amsta Rostfritt Supplier No. 5010550
  • ONLINE EXCLUSIVE PRICE
    1102.28 DKK
    949.00 DKK / Pakke med 1
    Spare 153.28 DKK
    13% Slukket
Forventet forsendelse: 13-01-2027
Highest Quality Stainless Steel lid For 608/609-150 Amsta transport container. Manufactured in Sweden.
  • Stainless steel 304 /1.4301
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel