Learn More
Stainless Steel gauze, 80 mesh woven from 0.14mm (0.0055in) dia wire, Type 316, Thermo Scientific Chemicals
233.99 DKK - 1147.62 DKK
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15487255
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046574.FL
75 x 75 mm
233.99 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
15424055
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046574.GT
150 x 150 mm
362.92 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
15434055
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046574.HB
300 x 300 mm
1147.62 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Description
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.