Learn More
Stainless Steel gauze, 40 mesh woven from 0.25mm (0.01in) dia wire, Type 304, Thermo Scientific Chemicals
459.05 DKK - 2265.43 DKK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356848
|
Thermo Scientific Alfa Aesar
013478.RR |
10 x 10 cm |
459.05 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11346848
|
Thermo Scientific Alfa Aesar
013478.CH |
20 x 30 cm |
894.25 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11366848
|
Thermo Scientific Alfa Aesar
013478.RU |
30 x 60 cm |
2265.43 DKK
Each |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.