Få mere at vide
BD Microlance™ Nåle i rustfrit stål
Nålemåler:
16 G
18 G
19 G
20 G
21 G
22 G
23 G
25 G
26 G
27 G
30 G
Længde (engelsk) Nål:
0.375 in.
0,5 tommer
0.5 in.
0,625 tommer
0.625 in.
1,0 tommer.
1.0 in.
1.25 in.
1,5 tommer
1.5 in.
2,0 tommer.
2.0 in.
Diameter (metrisk) Udvendig:
0.29 mm
0.4 mm
0.45 mm
0.5 mm
0.6 mm
0.7 mm
0.8 mm
0.9 mm
1.1 mm
1.25 mm
1.6 mm
Farve:
Beige
Blå
Black
Blue
Brown
Grå
Grøn
Gul
Hvid
Orange
Pink
Yellow
Pakningsstørrelse:
Pakke med 100
Pakke med 5000
Beskrivelse
Unique slip angle and polishing method results in less patient discomfort at the injection site and less risk for punching.
- Hypodermic needles with thin walls for optimal flow
- Fits syringes connected to a Luer adapter or Luer Lock connector
- Color coded for quick recognition of gauge for easy selection of appropriate flow rate
- Sterile and individually wrapped for hygienic, single-use application
- Stainless steel needles with epoxy resin adhesives
Tekniske data
Tekniske data
| Materiale | Stainless Steel with Epoxy Resin Adhesives |
| Farve | Brown |
| Oversigt | 5,000 individually wrapped needles, normal cannula wall |
| Nålemåler | 26 G |
| Diameter (metrisk) Udvendig | 0.45 mm |
| Til brug med (applikation) | For single use only |
| Latex fri | Yes |
| Længde (engelsk) Nål | 0.375 in. |
| Produkttype | Needle |
| Nålespids stil | Bevel, Short Angle |
| Vis mere |