Learn More
Cole-Parmer™ Stainless Steel Double-ended Spoon Set
Double-ended spoons feature a solid rounded shaft that is comfortable to hold for long periods.
Brand:  Cole-Parmer™ 06261-00
Additional Details : Weight : 0.80000kg
Product Code. 11760079
  • 990.00 DKK / Pack of 3
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • Set contains three spoons:
  • 5-7/8 in. (14.9cm) long spoon with 0.5mL and 2.0mL bowls
  • 8-1/4 in. (21.0cm) long spoon with 1.6 mL and 5.0 mL bowls
  • 11-7/8 in. (30.2cm) long spoon with 2.0 mL and 8.0 mL bowls
Specifications
Specifications
0.5 mL, 1.6 mL, 2 mL, 5 mL, 8 mL
3 Double-ended Spoons
14.9, 21, 30.2 cm
Spoon Set