Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E2
Adjusted to Meet or Exceed OIML E2 Tolerances
306.00 DKK - 17140.00 DKK
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
16652342
|
Ohaus™
30518004 |
1 mg |
306.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16662342
|
Ohaus™
30518005 |
2 mg |
306.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16672342
|
Ohaus™
30518006 |
5 mg |
306.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16682342
|
Ohaus™
30518007 |
10 mg |
306.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16692342
|
Ohaus™
30518008 |
20 mg |
306.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16602352
|
Ohaus™
30518009 |
50 mg |
306.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16612352
|
Ohaus™
30518010 |
100 mg |
306.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16622352
|
Ohaus™
30518011 |
200 mg |
306.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16632352
|
Ohaus™
30518012 |
500 mg |
306.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16642352
|
Ohaus™
30518013 |
1 g |
605.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16652352
|
Ohaus™
30518014 |
2 g |
631.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16662352
|
Ohaus™
30518015 |
5 g |
673.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16672352
|
Ohaus™
30518016 |
10 g |
695.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16682352
|
Ohaus™
30518017 |
20 g |
805.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16692352
|
Ohaus™
30518018 |
50 g |
930.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16602362
|
Ohaus™
30518019 |
100 g |
994.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16612362
|
Ohaus™
30518020 |
200 g |
1250.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
16622362
|
Ohaus™
30518021 |
500 g |
2100.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16632362
|
Ohaus™
30518022 |
1 kg |
3030.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16642362
|
Ohaus™
30518023 |
2 kg |
4860.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16652362
|
Ohaus™
30518024 |
5 kg |
7085.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16662362
|
Ohaus™
30518025 |
10 kg |
10255.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
16672362
|
Ohaus™
30518026 |
20 kg |
17140.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
Description
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
No Certificate | |
Protective Case |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ohaus™ Stainless Steel Calibration Weights - OIML E2