Learn More
Cole-Parmer™ Stainless Steel Bulkhead Union Compression Fitting
Brand:  Cole-Parmer™ 31406-13
Additional Details : Weight : 0.80000kg
Product Code. 11865443
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Bulkhead Union Compression Fitting
Fits 1/4 in. Tubing O.D.
1 Pack
Stainless Steel
-40°C
93°C