Learn More
Sampling Systems™ Stainless Steel Buckets with Skirt Band and Graduations
Sampling Systems™ Stainless Steel Buckets with Skirt Band and Graduations are made from heavy gauge, durable stainless steel and are ideal for hygienic applications.
Brand: Sampling Systems™ A264-12
Additional Details : Weight : 5.57380kg
Description
- Crevice free interior
- Graduated
- Supplied with skirt band
- Robust design
- 304 Stainless Steel
Specifications
12 L | |
Stainless Steel (304) | |
315 mm |
Yes | |
330 mm | |
Skirt Band |