Learn More
Bochem™ Stainless Steel Bowl
Brand:  Bochem™ 8735
Additional Details : Weight : 0.80000kg
Product Code. 10779431
  • 565.00 DKK / Each
Estimated Shipment: 28-05-2024
to see stock.
Specifications
250 x 180 x 70 mm
250 mm
Tray
1 Each
70 mm
Stainless Steel
180 mm
Rectangular