Learn More
Memmert™ Stacking Set for Stacking of Two Units of Same Size
Stacking Set for stacking of appliances of same size with subframe
Brand:  Memmert™ ICO150B42114
1708.56 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17199944
  • 1880.00 DKK / Each
Only null left
Estimated Shipment: 03-10-2025
Description
  • Only in connection with subframe B33509 or B43598
  • Consisting of stacking corners
  • One connecting plate for the rear, two wall brackets
Specifications
Specifications
ICO150
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Stacking Set for Stacking of Two Units of Same Size > ICO150