Få mere at vide
Thermo Scientific™ Stability Chamber, 311 L, Stainless Steel 1.4301, 2B
Temperature Range: 0°C to 60°C; Relative Humidity (RH): <95% at 37°C; Light: No Lighting
Brand: Thermo Scientific™ 3907
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- Optimum uniformity and a wide temperature range designed for easy compliance with regulatory requirements.
- Rugged construction and durable shelving for long life.
- Broad range of options to create the perfect fit for your applications.