Få mere at vide
Tocris Bioscience™ st-Ht31
Inhibits PKA/AKAP interactions; cell permeable
Brand: Tocris Bioscience™ 6286/1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
st-Ht31 is a stearated form of the peptide Ht-31. Inhibits the interaction between the RII subunits of cAMP-dependent PKA and A-kinase anchoring protein (AKAP) in cell extracts. Cell permeable.Tekniske data
| 188425-80-1 | |
| Store at -20°C | |
| 2798.27 | |
| 1 mg |