Få mere at vide
Tocris Bioscience™ SSYA10-001
Beskrivelse
SSYA10-001 is a potent coronavirus helicase inhibitor (IC50 values are 0.046 μM and >3 μM for unwinding and ATPase, respectively). Inhibits SARS-CoV-2 Nsp13 helicase (IC50 = 3.5 μM), blocks dsDNA and dsRNA unwinding activities of Nsp13 (IC50 values are 5.3 μM and 5.7 μM respectively). In a SARS-CoV replicon assay SSYA10-001 inhibits (+) sense but not (-) sense RNA synthesis (EC50 = 8.95 μM). Also inhibits human papillomavirus E6 (IC50 = 10 μM)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2,4-Dihydro-5-[[(2-nitrophenyl)thio]methyl]-4-(2-propen-1-yl)-3 H-1,2,4-triazole-3-thione |
| CAS | 675104-49-1 |
| Mængde | 10 mg |
| Mål | Antiviral Compounds |
| Molekylær formel | C12H12N4O2S2 |
| Renhed | 0.98 |