missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ SSR 180711 hydrochloride

Beskrivelse
SSR 180711 hydrochloride is a partial agonist at α7 nAChR. Enhances episodic memory and reverses MK-801 -induced deficits in retention of episodic memory. Increases extracellular dopamine levels in rat prefrontal cortex. Also exhibits antidepressant-like properties.
Specifications
Specifications
| Kemisk navn eller materiale | 4-Bromophenyl 1,4-diazabicyclo[3.2.2]nonane-4-carboxylate hydrochloride |
| CAS | 446031-79-4 |
| Mængde | 50 mg |
| Mål | Nicotinic (a7) Receptor Agonists |
| Molekylær formel | C14H17BrN2O2.HCl |
| Formel vægt | 361.66 |
| Renhed | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?