Få mere at vide
Tocris Bioscience™ SSM 3 trifluoroacetate
Potent furin inhibitor
Brand: Tocris Bioscience™ 5253/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SSM 3 trifluoroacetate is a potent furin inhibitor (EC50 = 54 nM). Blocks furin-dependent cell surface processing of anthrax protective antigen-83 in vitro.Tekniske data
| rel-N,N′′-[[(1R,3S,4S,6R)-4,6-Bis[(aminoiminomethyl)amino]-1,3-cyclohexanedyil]bis(oxy-4,1-phenylene)]bisguanidine tetratrifluoroacetate salt | |
| 922732-52-3 | |
| 10 mg |