Learn More
SSC Easy Sol 20X, MP Biomedicals™
Product Code. 11487140 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
5L
Unit Size:
5 L
Product Code. Quantity Unit Size
11487140 5L 5 L
1 options
Product Code. 11487140 Supplier MP Biomedicals Supplier No. 11SSC20X01
  • ONLINE EXCLUSIVE PRICE
    2240.00 DKK
    2131.37 DKK / 5 L
    Spare 108.63 DKK
    4% Slukket
 Restricted Item
Forventet forsendelse: 11-12-2026
SSC Easy Sol 20X, Molecular biology grade, Clear, Colorless Solution
  • SSC Buffer is used for preparing SSC buffer for Southern and Northen blotting methods.
  • The SSC buffer is utilized as buffer for DNA and RNA probe-containing hybridization cocktails
Specifications
Chemical Name or Material 3 M Trisodium citrate
Concentration 20X
pH 7.0 ±0.1
Physical Form Solution
Quantity 5L
DNase RNAse, DNAse, Protease free
Grade Molecular Biology