missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SS18L1 Antibody (7A7) [mFluor Violet 500 SE], Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42947MFV500
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SS18L1 Monoclonal antibody specifically detects SS18L1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Tekniske data
| SS18L1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| calcium-responsive transactivator, CRESTMGC78386, KIAA0693MGC26711, LP2261, SS18-like protein 1, synovial sarcoma translocation gene on chromosome 18-like 1, SYT homolog 1 | |
| synovial sarcoma translocation gene on chromosome 18-like 1 - (Uniprot# O75177) | |
| 0.1 mL | |
| Cancer | |
| 26039 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| 7A7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion