missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRPK2 Recombinant Human Protein
SRPK2, along with SRPK1, is most likely the cellular protein kinase mediating HBV core protein phosphorylation during viral infection.
13290.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_872633 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 105kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478593
|
Invitrogen™
PR7462A |
100μg |
13290.00 DKK
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10432133
|
Invitrogen™
PV4060 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SRPK2 represents an important host cell target for therapeutic intervention in HBV infection.
Specifications
NP_872633 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SRPK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
105kDa | |
RUO - research use only | |
SRPK2, SFRSK2 | |
Human | |
GST Tag | |
Frozen |