Få mere at vide
SRPIN340, MedChemExpress
Artikelnummer. 16456366
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16456366 100 mg -
16455928 10 mg 10mg
16442077 50 mg -
16408567 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16456366 Leverandør Medchem Express Leverandørnr. HY13949
MedChemExpress SRPIN340 is an ATP-competitive serine-arginine-rich protein kinase (SRPK) inhibitor, with a Ki of 0.89 μM for SRPK1.
SRPIN340 is an ATP-competitive serine-arginine-rich protein kinase (SRPK) inhibitor, with a Ki of 0.89 μM for SRPK1.
Tekniske data
Kemisk navn eller materiale SRPIN340
CAS 218156-96-8
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C18H18F3N3O
Mængde 100 mg
Synonym SRPK inhibitor
Opløselighedsinformation DMSO : ≥ 42 mg/mL (120.22 mM)
SMIL O=C(NC1=CC(C(F)(F)F)=CC=C1N2CCCCC2)C3=CC=NC=C3
Molekylvægt (g/mol) 349.35
Formel vægt 349.35
Procent renhed 98.64%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease