Få mere at vide
TERUMO SR STRAIGHT PUR VERSATUS™ Intravenous Catheters
Beskrivelse
- The Versatus™ I.V. Catheters are made out of medical grade polyurethane which becomes soft at body temperature ensuring patient comfort
- Precisely tapered and siliconised catheter for smooth advancement of the catheter
- Catheter with X-ray contrast strips for higher patient safety when placement goes wrong
- Made out of kink resistant material to prevent kinking during placement for a higher patient comfort
- Less chance of mechanical phlebitis because it is made out of high quality polyurethane so that it becomes softer at body temperature
Tekniske data
Tekniske data
| Materiale | Polyurethane, Stainless Steel (Needle) |
| Farve | Pink |
| Nålemåler | 20 G |
| Latex fri | Yes |
| Længde (metrisk) | 32 mm |
| Produkttype | IV Catheter |
| Engangs | Single-use |
| Til brug med (udstyr) | IV Lines |
| Tilslutningstype | Leur Slip |
| Uddannet | No |
| Vis mere |