Få mere at vide
TERUMO SR STRAIGHT FEP Intravenous Catheters SURFLO™
Artikelnummer. 15622714
Skift visning
Nålemåler:
20 G
24 G
Længde (engelsk) Nål:
0.75 in.
1.25 in.
2 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Nålemåler Længde (engelsk) Nål
15622714 24 G 0.75 in.
11867492 20 G 2 in.
11857492 20 G 1.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 15622714 Leverandør TERUMO Leverandørnr. 8SROX2419C1
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Short PIVC with an ETFE catheter tube and stainless steel internal needle
  • The Surflo I.V. Catheters are made out of medical grade EFTE
  • The materials of the Surflo™ I.V. Catheters are carefully selected to ensure a stable form during punctures under difficult conditions
  • Short PIVC with an ETFE catheter tube and stainless steel internal needle
  • Precisely tapered and siliconised catheter for smooth advancement of the catheter
  • Catheter with X-ray contrast strips for higher patient safety when placement goes wrong
  • The flashback chamber confirms needle tip entry into the vessel
  • The large range in catheters and related obturators allows most appropriate choice of product for a wide range of applications
TRUSTED_SUSTAINABILITY
Tekniske data
Materiale ETFE, Stainless Steel (Needle)
Farve Yellow
Nålemåler 24 G
Latex fri Yes
Længde (engelsk) Nål 0.75 in.
Længde (metrisk) 19 mm
Produkttype IV Catheter
Engangs Single-use
Til brug med (udstyr) IV Lines
Tilslutningstype Leur Slip
Uddannet No
Sterilitet Sterile