Få mere at vide
Tocris Bioscience™ SR 9238
Beskrivelse
SR 9238 is a potent and selective LXR inverse agonist (IC50 values are 43 and 214 nM for LXRβ and LXRα, respectively). Selectively inhibits liver LXR over peripheral LXR. Exhibits selectivity for LXR over a panel of 20 other nuclear receptors, including FXR. Reduces hepatic steatosis in obese mice.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 5-[[[[3′-(Methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl][(2,4,6-trimethylphenyl)sulfonyl]amino]methyl]-2-furancarboxylate |
| CAS | 1416153-62-2 |
| Mængde | 10 mg |
| Mål | LXR-like Receptor Antagonists |
| Molekylær formel | C31H33NO7S2 |
| Formel vægt | 595.73 |
| Renhed | >98% |