Få mere at vide
Tocris Bioscience™ SR 9238
Potent and selective LXR inverse agonist
Brand: Tocris Bioscience™ 5854/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SR 9238 is a potent and selective LXR inverse agonist (IC50 values are 43 and 214 nM for LXRβ and LXRα, respectively). Selectively inhibits liver LXR over peripheral LXR. Exhibits selectivity for LXR over a panel of 20 other nuclear receptors, including FXR. Reduces hepatic steatosis in obese mice.Tekniske data
| Ethyl 5-[[[[3′-(Methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl][(2,4,6-trimethylphenyl)sulfonyl]amino]methyl]-2-furancarboxylate | |
| 1416153-62-2 | |
| 50 mg |