Få mere at vide
Tocris Bioscience™ SR 58611A hydrochloride
GREENER_CHOICE
Artikelnummer. 18053892
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18053892 10 mg
18095842 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18053892 Leverandør Tocris Bioscience™ Leverandørnr. 3986/10
β3 agonist; displays anxiolytic and antidepressant effects
Selective β3-adrenergic receptor agonist. Displays both anxiolytic and antidepressant effects in rodent models. Orally active and brain penetrant.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 121524-09-2
Molekylær formel C22H27Cl2NO4
Tekniske data
Kemisk navn eller materiale SR 58611A hydrochloride
Mængde 10 mg
Formel vægt 440.36
Renhed >98%
Molekylvægt (g/mol) 440.361