Få mere at vide
Tocris Bioscience™ SR 48692
Beskrivelse
SR 48692 is a neurotensin antagonist; selective for NTS1 over NTS2 (apparent affinity, Ke, is 36 nM for NTS1). Competitively inhibits binding of [125I]-neurotensin to HT29 and N1E115 cell membranes (IC50 values are 15.3 and 20.4 nM respectively). Orally bioavailable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-[[[1-(7-Chloro-4-quinolinyl)-5-(2,6-dimethoxyphenyl)-1H-pyrazol-3-yl]carbonyl]amino]-tricyclo[3.3.1.13,7]decane-2-carboxylic acid |
| CAS | 146362-70-1 |
| Analyseprocentområde | >98% |
| Mængde | 5 mg |
| Molekylær formel | C32H31ClN4O5 |
| Formel vægt | Observed MW: 587.07 |