Få mere at vide
SR 16832, Tocris Bioscience™
PPAR gamma Receptor Antagonists
Brand: Tocris Bioscience 6383/25
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SR 16832 is a dual site PPARγ inhibitor. Acts at orthosteric and allosteric sites in the ligand binding domain. Inhibits binding of endogenous ligands and transcriptional activity of PPARγ, more effectively than the orthosteric covalent antagonists GW 9662 (Cat. No. 1508) and T 0070907 (Cat. No. 2301).Tekniske data
| 2088135-12-8 | |
| 2-Chloro-N-(6-methoxy-4-quinolinyl)-5-nitrobenzamide | |
| 25 mg |