Få mere at vide
Tocris Bioscience™ SR 13800
Potent MCT1 inhibitor
Brand: Tocris Bioscience™ 5431/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SR 13800 is a potent MCT1 inhibitor (IC50 = 0.5 nM); blocks proliferation of Raji lymphoma cells in vitro. Also inhibits lactate uptake in breast cancer cells in vitro.Tekniske data
| 2,6-Dihydro-7-[(3-hydroxypropyl)thio]-2-methyl-4-(2-methylpropyl)-6-(1-naphthalenylmethyl)-1H-pyrrolo[3,4-d]pyridazin-1one | |
| 227321-12-2 | |
| 50 mg |