Få mere at vide
Squalane-d62, TRC
Artikelnummer. 30418245
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30418245 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418245 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68375210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6,10,15,19,23-Hexamethyltetracosane-d62
CAS 16514-83-3
Renhedsgrad noter GC
Molekylær formel C30 D62
Mængde 10 mg
Synonym Tetracosane-1,1,1,2,3,3,4,4,5,5,6,7,7,8,8,9,9,10,11,11,12,12,13,13,14,14,15,16,16,17,17,18,18,19,20,20,21,21,22,22,23,24,24,24-d44','2,6,10,15,19,23-hexa(methyl-d3)- (8CI','9CI','ACI), 2,6,10,15,19,23-Hexa(methyl-d3)tetracosane-1,1,1,2,3,3,4,4,5,5,6,7,7,8,8,9,9,10,11,11,12,12,13,13,14,14,15,16,16,17,17,18,18,19,20,20,21,21,22,22,23,24,24,24-d44 (ACI), Perdeuteriosqualane, Squalane (C30D62), 2,6,10,15,19,23-Hexamethyltetracosane-d62, Squalane-D62
InChI formel InChI=1S/C30H62/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h25-30H,9-24H2,1-8H3/i1D3,2D3,3D3,4D3,5D3,6D3,7D3,8D3,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D,26D,27D,28D,29D,30D
SMIL [2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1,2,3,3,4,4,5,5,6,7,7,8,8,9,9,10,11,11,12,12,13,13,14,14,15,16,16,17,17,18,18,19,20,20,21,21,22,22,23,24,24,24-tetratetracontadeuterio-2,6,10,15,19,23-hexakis(trideuteriomethyl)tetracosane
Molekylvægt (g/mol) 485.1953
Formel vægt 484.8743
Procent renhed >95