Learn More
Squalane, 99%, Thermo Scientific Chemicals
Catalog Number p-4509606
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
111-01-3
PRAKJMSDJKAYCZ-UHFFFAOYSA-N
squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane
8089
2,6,10,15,19,23-hexamethyltetracosane
CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
111-01-3
PRAKJMSDJKAYCZ-UHFFFAOYSA-N
squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane
8089
2,6,10,15,19,23-hexamethyltetracosane
CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Acidity 0.2mg KOH/g max.
Melting Point 0.3°C
Color Colorless
Density 0.8085g/mL
Boiling Point 210°C to 215°C (1.0 mmHg)
Flash Point 217°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C30H62
Refractive Index 1.4510 to 1.4525
Quantity 100 mL
Saponification Value 1mg KOH/g max.
Beilstein 01,I,72
Merck Index 15,8895
Specific Gravity 0.8085
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in glacial acetic acid,readily soluble in ether,gasoline,benzene,,petroleum ether,chloroform and oils,slightly soluble in methanol,ethanol and acetone
Molecular Weight (g/mol) 422.82
Viscosity 48.2 mPa.s (20°C)
Formula Weight 422.82
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Squalane