missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ SPT Imidazopyridine 1

Beskrivelse
SPT Imidazopyridine 1 is a potent serine palmitoyl transferase (SPT) inhibitor (IC50 = 5 nM). Dose-dependently inhibits incorporation of 14C-serine into ceramide. Reduces plasma ceramide levels in vivo. Also increases plasma HDL and reduces vLDL cholesterol levels in rats.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[6-(1-Oxoheptyl)imidazo[1,2-a]pyridin-8-yl]benzeneacetic acid |
| CAS | 1933533-18-6 |
| Analyseprocentområde | >98% |
| Mængde | 50 mg |
| Molekylær formel | C22H24N2O3 |
| Formel vægt | Observed MW: 364.44 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?