missing translation for 'onlineSavingsMsg'
Få mere at vide

Spirotetramat-d5, TRC

Artikelnummer. 30419005
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419005 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419005 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68330225MG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Spirotetramat ethyl-d5
CAS 2469007-09-6
Renhedsgrad noter HPLC
Molekylær formel C21H22D5NO5
Mængde 25 mg
Synonym carbonic acid','cis-3-(2,5-dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro[4.5]dec-3-en-4-yl ethyl ester-d5
InChI formel InChI=1S/C21H27NO5/c1-5-26-20(24)27-18-17(16-12-13(2)6-7-14(16)3)19(23)22-21(18)10-8-15(25-4)9-11-21/h6-7,12,15H,5,8-11H2,1-4H3,(H,22,23)/t15-,21+/i1D3,5D2
SMIL O=C1C(C2=C(C)C=CC(C)=C2)=C(OC(OC([2H])([2H])C([2H])([2H])[2H])=O)[C@]3(CC[C@H](OC)CC3)N1
IUPAC navn (5s,8s)-3-(2,5-dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro[4.5]dec-3-en-4-yl (ethyl-d5) carbonate
Molekylvægt (g/mol) 378.48
Formel vægt 378.22
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.