Få mere at vide
Spirapril Hydrochloride, TRC
Artikelnummer. 30411832
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411832 100 mg
30427176 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30411832 Leverandør Toronto Research Chemicals Leverandørnr. TRCS682500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 94841-17-5
Molekylær formel C22 H30 N2 O5 S2 . Cl H
Molekylvægt (g/mol) 503.07
Synonym (8S)-7-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,4-dithia-7-azaspiro[4.4]nonane-8-carboxylic acid hydrochloride (1:1), [8S-[7[R*(R*)],8R*]]-7-[2-[[1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,4-dithia-7-azaspiro[4.4]nonane-8-carboxylic acid monohydrochloride, Renormax, Renpress, Sandopril, Spirapril hydrochloride, TI 211-950
IUPAC navn (8S)-7-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-1,4-dithia-7-azaspiro[4.4]nonane-8-carboxylic acid;hydrochloride
SMIL Cl.CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2CC3(C[C@H]2C(=O)O)SCCS3
Tekniske data
Kemisk navn eller materiale Spirapril hydrochloride
Mængde 100 mg
InChI formel InChI=1S/C22H30N2O5S2.ClH/c1-3-29-21(28)17(10-9-16-7-5-4-6-8-16)23-15(2)19(25)24-14-22(30-11-12-31-22)13-18(24)20(26)27;/h4-8,15,17-18,23H,3,9-14H2,1-2H3,(H,26,27);1H/t15-,17-,18-;/m0./s1
Formel vægt 502.1363