Learn More
Invitrogen™ SPHK1 Recombinant Human Protein
SPHK1, along with SPHK2, catalyzes the phosphorylation of sphingosine producing the signalling molecule sphingosine-1-phosphate (S1P).
Brand: Invitrogen™ PR8902A
Additional Details : Weight : 0.01000kg
Description
S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapuetics for the treatment of cancer.
Specifications
NP_068807.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
100μg | |
RUO - research use only | |
SPHK1 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
71.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SPHK and DGCK Kinase Family | |
Recombinant | |
Baculovirus |