missing translation for 'onlineSavingsMsg'
Få mere at vide

SPFH2 Antibody - BSA Free, R&D Systems™

Artikelnummer. 18609221 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.02 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18609221 0.02 mL
18693602 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18609221 Leverandør Novus Biologicals Leverandørnr. NBP2952420.02ml

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

SPFH2 Polyclonal antibody specifically detects SPFH2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPFH2
Applikationer Western Blot
Klassifikation Polyclonal
Konjugeret Unconjugated
Fortynding Western Blot 1:500-1:2000
Formulering PBS (pH 7.3), 50% glycerol
Gene Alias C8orf2SPFH domain-containing protein 2, Endoplasmic reticulum lipid raft-associated protein 2, ER lipid raft associated 2, Erlin-2, MGC87072, NET32, SPFH domain family, member 2, SPFH2chromosome 8 open reading frame 2, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-152 of human ERLIN2 (NP_001003790.1). KIEEGHIGVYYRGGALLTSTSGPGFHLMLPFITSYKSVQTTLQTDEVKNVPCGTSGGVMIYFDRIEVVNFLVPNAVYDIVKNYTADYDKALIFNKIHHELNQFCSVHTLQEVYIELFGLENDFSQESS
Oprensningsmetode Affinity purified
Mængde 0.02 mL
Regulatorisk status RUO
Forskningsdisciplin Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction
Primær eller sekundær Primary
Gen-id (Entrez) 11160
Målarter Human
Indhold og opbevaring Store at -20°C. Avoid freeze-thaw cycles.
Produkttype Antibody
Form Purified
Isotype IgG
Produktlinje Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.