Learn More
SPEX™ SamplePrep 6775 Freezer/Mill™ Accessories: Vial Set
For use with Spex SamplePrep Freezer/Mill Cryogenic Grinder
POA
Specifications
For Use With (Equipment) | Spex SamplePrep Freezer/Mill Cryogenic Grinder |
---|---|
Sample Volume (Metric) | 0.5 to 4 mL |
Description
Vial Set 6751
- Sample capacity: 0.5 to 4.0 mL
- Makes one complete vial plus three spare center sections
- Includes stainless-steel impactor, two end plugs, and four polycarbonate center cylinders (6751C4)
- Individual components available
Specifications
Spex SamplePrep Freezer/Mill Cryogenic Grinder | |
0.5 to 4 mL |