Learn More
3M™ Speedglas™ Welding Helmet 9100 with Side Windows
3M™ Speedglas™ Welding Helmet 9100 with Side Windows comes equipped with a 3M™ Speedglas™ Auto-darkening Filter 9100X.
Brand: 3M™ 7000000218
3220.00 DKK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- Variable dark shade 5, 8, 9-13 for most types of arc welding
- Wider field of view thanks to the side windows (shade 5 filters). Cover plates included as accessory
- Possible to lock-in any shade, e.g. in the light state 3 when grinding. Delay function to set the dark-to-light opening time
- Switching time (light-dark) 0.1 ms. The sensitivity of arc detection can be adjusted for unsurpassed switching reliability
- Tack welding comfort mode help minimize eye strain during extended tack welding applications
- Ergonomically designed head suspension
- Compatible with most 3M™ Safety Spectacles and/or 3M™ Disposable or Reusable Welding Particulate Respirators
Specifications
EN 175, EN 379, EN166:BT, EN175:B | |
Variable Dark Shade (5, 8, 9-13) | |
Auto Darkening Filter 9100X | |
Welding Helmet |