Få mere at vide
Spectrum™ Labs 3.5 - 5 kD Biotech RC Dialysis Kits
Artikelnummer. 15350782
Diameter (metrisk):
6.4 mm
Pakningsstørrelse:
0.5m
Artikelnummer. 15350782
Brand: Spectrum™ 133 192T
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
  • 0.5m Biotech RC tubing with glycerine
  • Two universal tubing closures
  • Five opening picks
With tubing made from Biotech RC membrane that can tolerate concentrated-weak acids, bases, dilute-strong acids, bases, most alcohols and some mild or dilute organic solvents
Tekniske data
MWCO 3.5 to 5 kDa
Diameter (metrisk) 6.4 mm
Længde (metrisk) 0.32 mL/cm vol./length
Mængde 0.5 m Tubing/Kit