missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin alpha 1 Antibody (SPTA1/2939R) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08346
Additional Details : Weight : 0.00970kg
Description
Spectrin alpha 1 Monoclonal specifically detects Spectrin alpha 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.Specifications
Spectrin alpha 1 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, ELISA : 2-4 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL | |
alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2) | |
Recombinant fragment of human Spectrin alpha 1 (around aa356-475) (exact sequence is proprietary) | |
100 μg | |
Cell Biology, Cellular Markers | |
6708 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
SPTA1/2939R | |
Unconjugated | |
10mM PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |