Få mere at vide
Molecular Devices SpectraMax™ iD3s MultiMode Platform STD with SoftMax Pro Software
Shop alle Molecular Devices produkterBeskrivelse
- Designed for multi-mode detection workflows in microplate-based assays
- Supports absorbance, fluorescence, and luminescence measurement applications
- Enables flexible assay setup with fully unable monochromator-based detection
- Supports biochemical and cell-based assay workflows across life science applications
- Provides a streamlined configuration for routine microplate reader workflows
- Includes preconfigured protocols to support common assay applications
- Integrated with SoftMax Pro Software for data acquisition and analysis
- Designed for ease of use and consistent results in laboratory environments
Tekniske data
Tekniske data
| Inkubator temperatur | 4-Zone Incubation to 70°C |
| Målehastighed | 11 sec. (96 wells), 22 sec. (384 wells) |
| Omrørere | Linear, Orbital, Double Orbital |
| Plade Type | 6 to 384-well Plates |
| Detektor type | PMT |
| Softwaretype | SoftMax Pro |
| Lyskilde | Xenon Flash |
| Læsemuligheder | Endpoint, Kinetic, Spectral Scanning |