Få mere at vide
Spectrum Labs™ Spectra/Por™ 1 membranskive
Diameter (metrisk):
47 mm
100 mm
Pakningsstørrelse:
Pakke med 50
Beskrivelse
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Tekniske data
Tekniske data
| Produkttype | Membrane Disc |
| MWCO | 6 to 8 kDa |
| membran | Regenerated Cellulose |
| Diameter (metrisk) | 100 mm |
| Til brug med (applikation) | For Basic Dialysis |